Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Amber Boston Round Bottles, Ultra Clean

General use bottles ideal for UV sensitive contents. Qorpak™ Amber Boston Round Bottles, Ultra Clean ensures product integrity for long-term storage.

Manufacturer:  Qorpak™ GLC05998

 View more versions of this product

Catalog No. 02-992-678


  Check Availability
Add to cart

Description

Description

General use bottles ideal for UV sensitive contents. Qorpak™ Amber Boston Round Bottles, Ultra Clean ensures product integrity for long-term storage.

  • Features a narrow screw neck opening, curved shoulders, and a round body
  • Glass offers minimal chemical interference with contents
  • Amber color absorb all wavelengths in the ultraviolet spectrum
  • Available with caps made of different materials (Black phenolic PolyCone and Green Thermoset F217 and PTFE lined)
Specifications

Specifications

16 oz., 480mL
16 oz.
F217 and PTFE
2.95 in.
Glass
Green
75mm
Screw
480mL
Thermoset
Amber
6.6 in.
12/Cs.
28-400
168mm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit