Learn More
Qorpak™ Amber Boston Round Bottles, Vacuum and Ionized: With Phenolic PolyCone Cap
General use bottles ideal for UV sensitive contents. Qorpak™ Amber Boston Round Bottles, Vacuum and Ionized: With Phenolic PolyCone Cap ensures product integrity for long-term storage.
Manufacturer:  Qorpak™ GLC01984
 View more versions of this product
Catalog No. 02-991-593
  Check Availability
Add to cart
Description
Description
Vacuum and Ionization Cleaning Process: Compressed air passes through a condensate separator and coalescing filter to remove aerosols and fine dust particles. Next, the air passes through an absorbent filter to remove gases. A vacuum draws the particulate-laden atmosphere out of the container. Individually cleaned closures are immediately placed on the container. Contaminants such as loose dirt, carton lint, aerosols, and fine glass particles are removed by the process.
Amber Boston Round Bottles
  • Narrow screw neck opening with curved shoulders
  • Glass offers minimal chemical interference with contents
Specifications
Specifications
16 oz., 480mL
16 oz.
Polycone
6.6 in.
Bulk pack
Black
2.95 in.
168mm
480mL
Phenolic
Amber
Glass
60/Cs. (Bulk pack)
28-400
75mm
With Black phenolic PolyCone lined cap