Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Amber Boston Round Bottles, Vacuum and Ionized: With Phenolic Pulp/Vinyl Cap

General use bottles ideal for UV sensitive contents. Qorpak™ Amber Boston Round Bottles, Vacuum and Ionized: With Phenolic Pulp/Vinyl Cap ensures product integrity for long-term storage.

Manufacturer:  Qorpak™ GLC01940

 View more versions of this product

Catalog No. 02-992-561


  Check Availability
Add to cart

Description

Description

  • Air inside the bottle is odor-free, tasteless and does not react with product
  • Contaminants such as loose dirt, carton lint, aerosols, and fine glass particles are removed by the Vacuum and Ionization process
  • Individually cleaned and sealed
  • Narrow screw neck opening with curved shoulders
  • Glass offers minimal chemical interference with contents
Specifications

Specifications

4 oz., 120mL
4 oz.
Pulp/Vinyl
4.4 in.
Convention pack
Black
1.88 in.
112mm
120mL
Phenolic
Amber
Glass
24/Cs.
22-400
48mm
Screw
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit