Learn More
Qorpak™ Amber Boston Round Bottles Without Cap Available on GSA/VA Contract for Federal Government customers only.
General use bottles ideal for UV sensitive contents. Qorpak™ Amber Boston Round Bottles Without Cap ensures product integrity for long-term storage.
Manufacturer:  Qorpak™ GLA00903
 View more versions of this product
Catalog No. 02-991-332
  Check Availability
Add to cart
Description
Description
  • Features a narrow screw neck opening, curved shoulders, and a round body
  • Glass offers minimal chemical interference with contents
  • Amber color absorb all wavelengths in the ultraviolet spectrum
  • This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
32 oz., 1000mL
32 oz.
3.78 in.
Glass
33-430
217mm
1000mL
Amber
8.5 in.
12/Cs.
96mm
Screw