Learn More
Qorpak™ Amber Boston Round Bottles Without Cap Available on GSA/VA Contract for Federal Government customers only.
General use bottles ideal for UV sensitive contents. Qorpak™ Amber Boston Round Bottles Without Cap ensures product integrity for long-term storage.
Manufacturer:  Qorpak™ GLA07052
 View more versions of this product
Catalog No. 02-991-287
  Check Availability
Add to cart
Description
Description
  • Features a narrow screw neck opening, curved shoulders, and a round body
  • Glass offers minimal chemical interference with contents
  • Amber color absorb all wavelengths in the ultraviolet spectrum
  • This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
16 oz., 500mL
16 oz.
2.9 in.
Glass
38-439
196mm
500mL
Amber
7.7 in.
24/Cs.
76mm
Screw