Learn More
Qorpak™ Amber Glass Jugs, Ultra Clean Available on GSA/VA Contract for Federal Government customers only.
Ideal for use when particulate contamination free containers are required for general lab use or fluid sampling. Qorpak™ Amber Glass Jugs, Ultra Clean ensures product integrity for long-term storage.
Manufacturer:  Qorpak™ GLC06805
 View more versions of this product
Catalog No. 02-992-304
  Check Availability
Add to cart
Description
Description
  • Ideal for light-sensitive products to protect against UV radiations
  • Perfect for storage of liquids in large quantity
  • Molded finger grips for easy handling
  • With phenolic F217/PTFE or thermoset F217/PTFE caps
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
128 oz., 3785mL
128 oz.
Thermoset
Amber
11.97 in.
4/Cs.
38-400
304mm
3785mL
Ultra-cleaned
F217 and PTFE
6.5 in.
Glass
Green
165 mm
Screw