Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Qorpak™ Amber Glass Jugs, Vacuum and Ionized - Phenolic F217 and PTFE Caps Available on GSA/VA Contract for Federal Government customers only.

Ideal for light-sensitive products to protect from harmful light and UV-rays

Manufacturer:  Qorpak™ GLC07021

 View more versions of this product

Catalog No. 02-992-551



Description

Description

  • Molded finger grips facilitates easy handling
  • Contaminants such as loose dirt, carton lint, aerosols, and fine glass particles are removed by Vacuum and Ionization process
  • Air inside the jug is odor-free, tasteless and does not react with product
  • Separate washing not required, thereby saving time
  • Final finished product kept clean and dust-free

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

84 oz., 2500mL
84 oz.
Phenolic
Amber
12.2 in.
6/Cs.
38-430
310mm
Screw
2500mL
Vacuum and ionized
F217 and PTFE
5.39 in.
Glass
Black
137 mm
Narrow
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit