Learn More
Qorpak™ Amber Vials with Black Phenolic Lined Caps Available on GSA/VA Contract for Federal Government customers only.
Exceptionally resistant to heat shock and chemical leaching
$168.00 - $251.00
Specifications
Closure Type Screw Cap
For Use With (Application) Provides protection against harmful light and UV rays
View More Specs
Products 6
Catalog Number Mfr. No. Volume (Metric) Volume (English) Height (English) Liner Material Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Volume (English) Height (English) Liner Material Price Quantity & Availability  
02-991-364
View Documents
Qorpak™
GLC10521
3.75 mL 1 dr. 1.77 in. Rubber
Case of 144 for $168.00
Add to cart
02-993-212 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Qorpak™
GLC07411
3.75 mL 1 dr. 1.77 in. Polycone
Case of 144 for $251.00
Add to cart
02-993-223
View Documents
Qorpak™
GLC11283
1.85 mL 0.5 dr. 1.37 in. Pulp/Vinyl
Case of 144 for $217.00
Add to cart
02-993-237 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Qorpak™
GLC11281
15 mL 4 dr. 2.75 in. Pulp/Vinyl
Case of 144 for $251.00
Add to cart
02-993-279
View Documents
Qorpak™
GLC11282
7.5 mL 2 dr. 2.36 in. Pulp/Vinyl
Case of 144 for $236.00
Add to cart
02-993-335 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Qorpak™
GLC11284
7.5 mL 2 dr. 2.36 in. Pulp/Vinyl
Case of 144 for $194.00
Add to cart
Description
Description
  • Made of Type I borosilicate glass
  • Provides protection against harmful light and UV rays
  • Available with different cap materials (Phenolic PolyCone, Phenolic Pulp/Vinyl and Phenolic Rubber)
Amber Vials with Pulp/Vinyl Lined Caps
  • High density polyethylene coated paper and laminated to pulpboard
  • Used for food, beverage, medical and chemical applications
  • Packed at <120°F
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Screw Cap
Provides protection against harmful light and UV rays