Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Amber Wide Mouth Packer Bottles with Green Thermoset F217 and PTFE Cap

General use bottles ideal for UV sensitive contents. Qorpak™ Amber Glass Jug with Green Thermoset F217 and PTFE Cap are used in chemical and environmental applications.

Manufacturer:  Qorpak™ GLC02132

 View more versions of this product

Catalog No. 02-993-006


  Check Availability
Add to cart

Description

Description

  • Feature a wide opening for making them the ideal general purpose bottle for solids
  • Classic "vitamin bottles" and are often used in pharmaceutical packaging
  • Closure shows high temperature tolerance with wide range of chemical compatibility
Specifications

Specifications

17 oz., 500mL
17 oz.
F217 and PTFE
3.19 in.
Glass
Green
81mm
Screw
500mL
Thermoset
Amber
5.75 in.
60/Cs.
53-400
146mm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit