Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Amber Wide Mouth Packer Bottles, Ultra Clean Available on GSA/VA Contract for Federal Government customers only.

Designed for general lab use or fluid sampling. Qorpak™ Amber Wide Mouth Packer Bottles, Ultra Clean provides additional protection for light-sensitive contents by blocking UV rays.

Manufacturer:  Qorpak™ GLC06407

 View more versions of this product

Catalog No. 02-992-299


  Check Availability
Add to cart

Description

Description

  • Wide opening for easy access to content
  • Ideal for sampling of solids
  • Glass offers minimal chemical interference with contents
  • Useful in wide range of applications including pharmaceutical, chemical and environmental

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

84 oz., 2500mL
84 oz.
F217 and PTFE
5.51 in.
Glass
Green
140mm
Screw
2500mL
Thermoset
Amber
9.41 in.
12/Cs.
70-400
239mm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit