Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Amber Wide Mouth Packer Bottles, Ultra Clean

Designed for general lab use or fluid sampling. Qorpak™ Amber Wide Mouth Packer Bottles, Ultra Clean provides additional protection for light-sensitive contents by blocking UV rays.

Manufacturer:  Qorpak™ GLC07129

 View more versions of this product

Catalog No. 02-992-951


  Check Availability
Add to cart

Description

Description

  • Wide opening for easy access to content
  • Ideal for sampling of solids
  • Glass offers minimal chemical interference with contents
  • Useful in wide range of applications including pharmaceutical, chemical and environmental
Specifications

Specifications

1 oz., 30mL
1 oz.
F217 and PTFE
1.46 in.
Glass
Green
37mm
Screw
30mL
Thermoset
Amber
2.56 in.
24/Cs.
28-400
65mm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit