Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Amber Wide Mouth Packer Bottles with Black Phenolic Solid PE Cap

General use bottles ideal for UV sensitive contents. Qorpak™ Amber Glass Jug with Black Phenolic Solid PE Cap are used in chemical and environmental applications.

Manufacturer:  Qorpak™ GLC02102

 View more versions of this product

Catalog No. 02-992-611


  Check Availability
Add to cart

Description

Description

  • Feature a wide opening for making them the ideal general purpose bottle for solids
  • Classic "vitamin bottles" and are often used in pharmaceutical packaging
Specifications

Specifications

2 oz., 60mL
2 oz.
Polyethylene
1.73 in.
Glass
216/Cs.
33-400
75mm
60mL
Phenolic
Amber
2.95 in.
Bulk pack
Black
44mm
Screw
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit