Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Amber Wide Mouth Packer Bottles without Cap

General use bottles ideal for UV sensitive contents. Qorpak™ Amber Wide Mouth Packer Bottles without Cap is used in chemical and environmental applications.

Manufacturer:  Qorpak™ GLA00928

 View more versions of this product

Catalog No. 02-992-396


  Check Availability
Add to cart

Description

Description

  • Feature a wide opening, making them the ideal general purpose bottle for solids
  • Classic "vitamin bottles" and are often used in pharmaceutical packaging
Specifications

Specifications

32 oz., 950mL
32 oz.
3.9 in.
Glass
12/Cs.
99mm
Screw
950mL
Amber
7.01 in.
Qorpak Carton
53-400
178mm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit