Learn More
Qorpak™ Black Phenolic 0.030 Solid Polyethylene Lined Caps Available on GSA/VA Contract for Federal Government customers only.
Ideal for nail polish remover and higher concentration acids and alkalis. Qorpak™ Black Phenolic 0.030 Solid Polyethylene Lined Caps offer good chemical resistance and low moisture vapor transmission rate.
Manufacturer:  Qorpak™ CAP00091
 View more versions of this product
Catalog No. 02-993-288
  Check Availability
Add to cart
Description
Description
  • Paperless composition avoids contamination from fiber or paper particles
  • For non-oil products filled at room temperature
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Polyethylene
144
Bagged
89-400
Black
12/Pk.
Wide mouth bottle
Smooth