Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Qorpak™ Black Phenolic 0.030 Solid Polyethylene Lined Caps
Ideal for nail polish remover and higher concentration acids and alkalis. Qorpak™ Black Phenolic 0.030 Solid Polyethylene Lined Caps offer good chemical resistance and low moisture vapor transmission rate.
Manufacturer:  Qorpak™ CAP00286
 View more versions of this product
Catalog No. 02-991-035
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Paperless composition avoids contamination from fiber or paper particles
  • For non-oil products filled at room temperature
Specifications
Specifications
Polyethylene
910
Wide mouth bottle
Smooth
Black
Bulk
58-400