Qorpak™ Black Phenolic 0.030 Solid Polyethylene Lined Caps Available on GSA/VA Contract for Federal Government customers only.

Ideal for nail polish remover and higher concentration acids and alkalis. Qorpak™ Black Phenolic 0.030 Solid Polyethylene Lined Caps offer good chemical resistance and low moisture vapor transmission rate.

Manufacturer: Qorpak™ CAP00308

 View more versions of this product

Catalog No. 02-991-052

  Check Availability
Add to Cart

    Specifications

    No. per Case 598
    Closure Liner Polyethylene
    Closure Type Smooth
    Color Black
    For Use With Wide mouth bottle
    Neck Finish 70-400
    Packaging Bulk

    • Paperless composition avoids contamination from fiber or paper particles
    • For non-oil products filled at room temperature

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit