Learn More
Qorpak™ Black Phenolic 0.030 Solid Polyethylene Lined Caps Available on GSA/VA Contract for Federal Government customers only.
Ideal for nail polish remover and higher concentration acids and alkalis. Qorpak™ Black Phenolic 0.030 Solid Polyethylene Lined Caps offer good chemical resistance and low moisture vapor transmission rate.
Manufacturer:  Qorpak™ CAP00243
 View more versions of this product
Catalog No. 02-991-123
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications
Description
  • Paperless composition avoids contamination from fiber or paper particles
  • For non-oil products filled at room temperature
Description & Specifications
Specifications
Polyethylene
1638
Wide mouth bottle
Smooth
Black
Bulk
43-400