Learn More
Qorpak™ Black Phenolic F217 and PTFE Lined Caps Available on GSA/VA Contract for Federal Government customers only.
Ideal for use with a wide range of chemicals including analytical lab samples, high purity chemicals, strong acids solvents, environmental samples, pharmaceuticals and diagnostic reagents. Qorpak™ Black Phenolic F217 and PTFE Lined Caps offer good chemical and temperature resistance.
Manufacturer:  Qorpak™ CAP06689
 View more versions of this product
Catalog No. 02-993-402
  Check Availability
Add to cart
Description
Description
  • Adherence consistency to close dimensional tolerances
  • Resist from chemical attacks at room temperature
  • Provide resilient tight seal with low moisture transmission rate
  • Available in ribbed or smooth style
  • Available in bagged or bulk packaging
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
F217 and PTFE
576
Bagged
20-400
Black
12/Pk.
Wide mouth bottle
Ribbed