Learn More
Qorpak™ Black Phenolic F217 and PTFE Lined Caps Available on GSA/VA Contract for Federal Government customers only.
Ideal for use with a wide range of chemicals including analytical lab samples, high purity chemicals, strong acids solvents, environmental samples, pharmaceuticals and diagnostic reagents. Qorpak™ Black Phenolic F217 and PTFE Lined Caps offer good chemical and temperature resistance.
Manufacturer:  Qorpak™ CAP06694
 View more versions of this product
Catalog No. 02-993-428
  Check Availability
Add to cart
Specifications
Description
  • Adherence consistency to close dimensional tolerances
  • Resist from chemical attacks at room temperature
  • Provide resilient tight seal with low moisture transmission rate
  • Available in ribbed or smooth style
  • Available in bagged or bulk packaging
Description & Specifications
Specifications
F217 and PTFE
576
Bagged
38-400
Black
12/Pk.
Wide mouth bottle
Ribbed