Qorpak™ Black Phenolic F217 and PTFE Lined Caps Available on GSA/VA Contract for Federal Government customers only.

Ideal for use with a wide range of chemicals including analytical lab samples, high purity chemicals, strong acids solvents, environmental samples, pharmaceuticals and diagnostic reagents. Qorpak™ Black Phenolic F217 and PTFE Lined Caps offer good chemical and temperature resistance.

Manufacturer: Qorpak™ CAP00222

 View more versions of this product

Catalog No. 02-991-042

  Check Availability
Add to Cart

    Specifications

    No. per Case 1100
    Closure Liner F217 and PTFE
    Closure Type Ribbed
    Color Black
    For Use With Bottle
    Neck Finish 33-430
    Packaging Bulk

    • Adherence consistency to close dimensional tolerances
    • Resist from chemical attacks at room temperature
    • Provide resilient tight seal with low moisture transmission rate
    • Available in ribbed or smooth style
    • Available in bagged or bulk packaging

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit