Learn More
Qorpak™ Black Phenolic PolyCone Lined Caps Available on GSA/VA Contract for Federal Government customers only.
Forms an exceptionally tight seal and offer a good chemical barrier. Qorpak™ Black Phenolic PolyCone Lined Caps is used for everything from liquids, to soaps, to macro-invertebrates.
Manufacturer:  Qorpak™ CAP00170
 View more versions of this product
Catalog No. 02-991-077
  Check Availability
Add to cart
Description
Description
  • Molded liner around the sealing areas eliminates leakage, evaporation, contamination, binding and backing off
  • Available in ribbed style
  • Available in bulk/bagged packaging
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Polycone
4100 Cs.
Wide mouth bottle
Ribbed
Black
Bulk
24-400