Learn More
Qorpak™ Black Phenolic Pulp/Aluminum Foil Lined Caps Available on GSA/VA Contract for Federal Government customers only.
Ideal for non-acid and non-alkaline products. Qorpak™ Black Phenolic Pulp/Aluminum Foil Lined Caps are typically used for food applications.
Manufacturer:  Qorpak™ CAP00086
 View more versions of this product
Catalog No. 02-991-038
  Check Availability
Add to cart
Specifications
Description
  • Closure liners free of odor and taste
  • Used for organic solvents, chrome cleaners, brake fluids and mineral oils
  • Neck finish for narrow bottles is 425 GPI and 400 for wider
  • Available as ribbed style
  • Available in bulk/bagged packaging
Description & Specifications
Specifications
Pulp/Aluminum Foil
304 Cs.
Wide mouth bottle
Smooth
Black
Bulk
89-400