Learn More
Qorpak™ Black Phenolic Pulp/Aluminum Foil Lined Caps Available on GSA/VA Contract for Federal Government customers only.
Ideal for non-acid and non-alkaline products. Qorpak™ Black Phenolic Pulp/Aluminum Foil Lined Caps are typically used for food applications.
Manufacturer:  Qorpak™ CAP00124
 View more versions of this product
Catalog No. 02-993-248
  Check Availability
Add to cart
Specifications
Description
  • Closure liners free of odor and taste
  • Used for organic solvents, chrome cleaners, brake fluids and mineral oils
  • Neck finish for narrow bottles is 425 GPI and 400 for wider
  • Available as ribbed style
  • Available in bulk/bagged packaging
Description & Specifications
Specifications
Pulp/Aluminum Foil
576 Cs.
Bagged
15-425
Black
12/Pk.
Narrow mouth bottle
Ribbed