Qorpak™ Black Phenolic Pulp/Aluminum Foil Lined Caps Available on GSA/VA Contract for Federal Government customers only.

Ideal for non-acid and non-alkaline products. Qorpak™ Black Phenolic Pulp/Aluminum Foil Lined Caps are typically used for food applications.

Manufacturer: Qorpak™ CAP00206

 View more versions of this product

Catalog No. 02-991-079

  Check Availability
Add to Cart

    Specifications

    Closure Liner Pulp/Aluminum Foil
    Closure Type Smooth
    Color Black
    For Use With Wide mouth bottle
    Neck Finish 33-400
    No. per Case 2400 Cs.
    Packaging Bulk

    • Closure liners free of odor and taste
    • Used for organic solvents, chrome cleaners, brake fluids and mineral oils
    • Neck finish for narrow bottles is 425 GPI and 400 for wider
    • Available as ribbed style
    • Available in bulk/bagged packaging

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit