Learn More
Qorpak™ Black Phenolic Pulp/Tinfoil Lined Caps Available on GSA/VA Contract for Federal Government customers only.
For use with solids and powders. Qorpak™ Black Phenolic Pulp/Tinfoil Lined Caps offer strong resistance to chemicals and temperature.
Manufacturer:  Qorpak™ CAP00108
 View more versions of this product
Catalog No. 02-991-432
  Check Availability
Add to cart
Specifications
Description
  • Not recommended for acids and alkalis
  • Available as ribbed and smooth style
  • Thread finish for narrow bottles is 425 GPI and for wider 400
  • Available as bulk/bagged packaging
Description & Specifications
Specifications
Pulp/Tin Foil
13500 Cs.
Narrow mouth bottle
Ribbed
Black
Bulk
13-425