Qorpak™ Black Phenolic Pulp/Tinfoil Lined Caps Available on GSA/VA Contract for Federal Government customers only.

For use with solids and powders. Qorpak™ Black Phenolic Pulp/Tinfoil Lined Caps offer strong resistance to chemicals and temperature.

Manufacturer: Qorpak™ CAP00136

 View more versions of this product

Catalog No. 02-993-326

  Check Availability
Add to Cart

    Specifications

    Closure Liner Pulp/Tin Foil
    Closure Type Ribbed
    Color Black
    For Use With Wide mouth bottle
    Neck Finish 18-400
    No. per Case 576 Cs.
    No. per Pack 12/Pk.
    Packaging Bagged

    • Not recommended for acids and alkalis
    • Available as ribbed and smooth style
    • Thread finish for narrow bottles is 425 GPI and for wider 400
    • Available as bulk/bagged packaging

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit