Learn More
Qorpak™ Black Phenolic Pulp/Vinyl Lined Caps
Used for food, beverage, medical and chemical applications packed at <120°F. Qorpak™ Black Phenolic Pulp/Vinyl Lined Caps are ideal for general use.
Manufacturer:  Qorpak™ CAP00085
 View more versions of this product
Catalog No. 02-993-357
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • HDPE coated paper liner
  • Used for mild acids, alkalis, solvents, alcohols, oils and aqueous products
  • Not recommended for active hydrocarbons or bleaches
  • Available in smooth and ribbed style
  • Available as bulk/bagged packaging
Specifications
Specifications
Pulp/Vinyl
144 Cs.
Bagged
83-400
Black
12/Pk.
Wide mouth bottle
Smooth