Qorpak™ Black Phenolic Pulp/Vinyl Lined Caps Available on GSA/VA Contract for Federal Government customers only.

Ideal for general use

Manufacturer: Qorpak™ CAP-00082

 View all options for this product

Catalog No. 02-991-071

  Check Availability
Add to Cart


    Closure Liner Pulp/Vinyl
    Closure Type Smooth
    Color Black
    For Use With Wide mouth bottle
    Neck Finish 83-400
    No. per Case 340 Cs.
    Packaging Bulk

    • HDPE coated paper liner
    • Made of different materials (Phenolic Pulp/Vinyl and Polypropylene Pulp/Vinyl)
    • Used for food, beverage, medical and chemical applications packed at <120°F
    • Not recommended for active hydrocarbons or bleaches
    • Available in smooth and ribbed style
    • Available as bulk/bagged packaging

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit