Qorpak™ Black Phenolic Pulp/Vinyl Lined Caps Available on GSA/VA Contract for Federal Government customers only.

Used for food, beverage, medical and chemical applications packed at <120°F. Qorpak™ Black Phenolic Pulp/Vinyl Lined Caps are ideal for general use.

Manufacturer: Qorpak™ CAP00209

 View more versions of this product

Catalog No. 02-991-080

  Check Availability
Add to Cart

    Specifications

    Closure Liner Pulp/Vinyl
    Closure Type Smooth
    Color Black
    For Use With Wide mouth bottle
    Neck Finish 33-400
    No. per Case 2100 Cs.
    Packaging Bulk

    • HDPE coated paper liner
    • Used for mild acids, alkalis, solvents, alcohols, oils and aqueous products
    • Not recommended for active hydrocarbons or bleaches
    • Available in smooth and ribbed style
    • Available as bulk/bagged packaging

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit