Learn More
Qorpak™ Black Phenolic Pulp/Vinyl Lined Caps
Used for food, beverage, medical and chemical applications packed at <120°F. Qorpak™ Black Phenolic Pulp/Vinyl Lined Caps are ideal for general use.
Manufacturer:  Qorpak™ CAP00245
 View more versions of this product
Catalog No. 02-991-094
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • HDPE coated paper liner
  • Used for mild acids, alkalis, solvents, alcohols, oils and aqueous products
  • Not recommended for active hydrocarbons or bleaches
  • Available in smooth and ribbed style
  • Available as bulk/bagged packaging
Specifications
Specifications
Pulp/Vinyl
1760 Cs.
Wide mouth bottle
Smooth
Black
Bulk
43-400