Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Black Phenolic Rubber Lined Caps

Catalog Number 02993293
Click to view available options
Neck Finish:
13-425
15-425
18-400
20-400
22-400
24-400
24-414
28-400
33-400
38-400
43-400
48-400
No. per Case:
11000 Cs.
13500 Cs.
1400 Cs.
2160 Cs.
3000 Cs.
4000 Cs.
4600 Cs.
576 Cs.
Packaging:
Bagged
Bulk

Intented for repeated use. Qorpak™ Black Phenolic Rubber Lined Caps are autoclavable.

  • Liner exhibit resiliency
  • Resistant to moisture vapor
  • Provides consistent adherence to close dimensional tolerance
  • Available in ribbed and smooth style
  • Available as bulk/bagged packaging

Compliance

FDA 21 CFR177.2600

Closure Liner Rubber
Closure Type Ribbed
For Use With (Equipment) Narrow mouth bottle
Color Black
Material Phenolic Resin
No. per Case 576 Cs.
No. per Pack 12/Pk.
Packaging Bagged
Neck Finish 13-425
Certifications/Compliance FDA 21 CFR 177.2600
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit