Qorpak™ Black Phenolic Rubber Lined Caps Available on GSA/VA Contract for Federal Government customers only.

Intented for repeated use. Qorpak™ Black Phenolic Rubber Lined Caps are autoclavable.

Manufacturer: Qorpak™ CAP05842

 View more versions of this product

Catalog No. 02-993-374

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance FDA 21 CFR 177.2600
    Closure Liner Rubber
    Closure Type Smooth
    Color Black
    For Use With Wide mouth bottle
    Neck Finish 48-400
    No. per Case 576 Cs.
    No. per Pack 12/Pk.
    Packaging Bagged

    • Liner exhibit resiliency
    • Resistant to moisture vapor
    • Provides consistent adherence to close dimensional tolerance
    • Available in ribbed and smooth style
    • Available as bulk/bagged packaging

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit