Learn More
Qorpak™ Black Phenolic Rubber Lined Caps Available on GSA/VA Contract for Federal Government customers only.
Intented for repeated use. Qorpak™ Black Phenolic Rubber Lined Caps are autoclavable.
Manufacturer:  Qorpak™ CAP00480
 View more versions of this product
Catalog No. 02-991-140
  Check Availability
Add to cart
Description
Description
  • Liner exhibit resiliency
  • Resistant to moisture vapor
  • Provides consistent adherence to close dimensional tolerance
  • Available in ribbed and smooth style
  • Available as bulk/bagged packaging
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
FDA 21 CFR 177.2600
Black
Bulk
24-400
Rubber
4000 Cs.
Wide mouth bottle
Smooth