Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Qorpak™ Black Polypropylene Unlined Caps
Click to view available options
Neck Finish:
22-400
24-400
28-400
33-400
38-400
43-400
45-400
48-400
53-400
58-400
63-400
70-400
83-400
89-400
For Use With (Equipment):
Wide mouth bottle
No. per Case:
1300 Cs.
144 Cs.
1700 Cs.
2000 Cs.
2200 Cs.
2900 Cs.
4000 Cs.
500 Cs.
5000 Cs.
580 Cs.
6500 Cs.
760 Cs.
8500 Cs.
900 Cs.
Packaging:
Bagged
Bulk
Description
- Offers good impact strength and pliability
- Available in ribbed style
- Available as bulk/bagged packaging
Specifications
Closure Type | Ribbed |
For Use With (Equipment) | Wide mouth bottle |
Color | Black |
Material | Polypropylene |
No. per Case | 4000 Cs. |
Packaging | Bulk |
Neck Finish | 33-400 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction