Learn More
Qorpak™ Black Polypropylene Unlined Caps Available on GSA/VA Contract for Federal Government customers only.
Ideally used for dry products. Qorpak™ Black Polypropylene Unlined Caps are autoclavable.
Manufacturer:  Qorpak™ CAP00396
 View more versions of this product
Catalog No. 02-993-071
  Check Availability
Add to cart
Specifications
Description
  • Offers good impact strength and pliability
  • Available in ribbed style
  • Available as bulk/bagged packaging
Description & Specifications
Specifications
Black
100/Pk.
Wide mouth bottle
Ribbed
500 Cs.
Bagged
53-400