Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Black Polypropylene Unlined Caps

Catalog Number 02993149
Click to view available options
Neck Finish:
22-400
24-400
28-400
33-400
38-400
43-400
45-400
48-400
53-400
58-400
63-400
70-400
83-400
89-400
For Use With (Equipment):
Wide mouth bottle
No. per Case:
1300 Cs.
144 Cs.
1700 Cs.
2000 Cs.
2200 Cs.
2900 Cs.
4000 Cs.
500 Cs.
5000 Cs.
580 Cs.
6500 Cs.
760 Cs.
8500 Cs.
900 Cs.
Packaging:
Bagged
Bulk

Ideally used for dry products. Qorpak™ Black Polypropylene Unlined Caps are autoclavable.

  • Offers good impact strength and pliability
  • Available in ribbed style
  • Available as bulk/bagged packaging
Closure Type Ribbed
For Use With (Equipment) Wide mouth bottle
Color Black
Material Polypropylene
No. per Case 500 Cs.
No. per Pack 100/Pk.
Packaging Bagged
Neck Finish 45-400
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit