Learn More
Qorpak™ Black Polypropylene Unlined Caps Available on GSA/VA Contract for Federal Government customers only.
Ideally used for dry products. Qorpak™ Black Polypropylene Unlined Caps are autoclavable.
Manufacturer:  Qorpak™ CAP00377
 View more versions of this product
Catalog No. 02-993-304
  Check Availability
Add to cart
Description
Description
  • Offers good impact strength and pliability
  • Available in ribbed style
  • Available as bulk/bagged packaging
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Black
100/Pk.
Bottle
Ribbed
500 Cs.
Bagged
38-430