Learn More
Qorpak™ Tamper Evident Caps For HDPE Square Bottles Available on GSA/VA Contract for Federal Government customers only.
For HDPE square bottles. Qorpak™ Blue Tamper Evident Caps are made of polypropylene.
Manufacturer:  Qorpak™ 241718
 View more versions of this product
Catalog No. 02-994-007
  Check Availability
Add to cart
Description
Description
  • Caps have built in seal with integrated tamper evidence ring
  • Caps and bottles sold separately
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Polypropylene
2500/Cs.
Blue
DIN28