Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Qorpak™ Borosilicate Glass Crimp Neck Serum Vials without Caps
Ideal for pharmaceutical packaging, lyophilization, and sample storage
$506.50 - $819.00
Specifications
Material | Borosilicate Glass |
---|---|
For Use With (Application) | Ideal for pharmaceutical packaging, lyophilization, and sample storage |
Thread Style | Crimp |
Catalog Number | Mfr. No. | Volume (Metric) | Color | Volume (English) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Color | Volume (English) | Price | Quantity & Availability | |||
02-991-102
|
Qorpak™
235278 |
5 mL | Amber | 1.4 dr. |
Case of 1200 for $819.00
|
|
|||
02-991-196
|
Qorpak™
235281 |
10 mL | Amber | 2.7 dr. |
Case of 800 for $543.00
|
|
|||
02-991-244
|
Qorpak™
235282 |
10 mL | Clear | 2.7 dr. |
Case of 800 for $506.50
|
|
|||
02-991-290
|
Qorpak™
235279 |
5 mL | Clear | 1.4 dr. |
Case of 1200 for $751.00
|
|
|||
02-991-214
|
Qorpak™
235276 |
2 mL | Amber | 0.5 dr. | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
02-991-210
|
Qorpak™
235277 |
2 mL | Clear | 0.5 dr. | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
Description
- Break resistant and typically used for handling and containment of pharmaceuticals and serums
- Suitablel for packing and storing blood plasma, media, reagents and vaccines
- Available in two colors; clear for maximum visibility and amber for UV protection
Specifications
Borosilicate Glass | |
Ideal for pharmaceutical packaging, lyophilization, and sample storage | |
Crimp |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title