Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Clear Borosilicate Sample Vials with Caps Available on GSA/VA Contract for Federal Government customers only.

Highly resistant to heat shock and chemical leaching

Manufacturer:  Qorpak™ GLC04883

 View more versions of this product

Catalog No. 02-992-668


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Screw cap
10 dr., 40mL
29.25mm
10 dr.
Sample Vial
40mL
3.18 in.
Highly resistant to heat and chemicals
Screw
144/Cs.
40mL
1.15 in.
Clear
Borosilicate Glass
24mm-400
81mm
Phenolic
Pulp/Vinyl
10 dr.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit