Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Clear Borosilicate Vial with Phenolic Pulp/Tin Foil Lined Cap

Offers maximum visibility

Specifications

Closure Type Screw Cap
Color Clear
Material Borosilicate Glass
For Use With (Application) Provide resistance to heat shock and chemical leaching
Closure Material Phenolic
View More Specs

Products 2
Catalog Number Mfr. No. Volume (Metric) Quantity Volume (English) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Quantity Volume (English) Price Quantity & Availability  
02-992-612
View Documents
Qorpak™
GLC01036
40 mL 10 dr. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
02-992-641
View Documents
Qorpak™
GLC01037
40 mL 10 dr. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

  • Material: Type I Borosilicate glass
  • Provide resistance to heat shock and chemical leaching
  • General use liner: commonly used for food, beverage, medical and chemical applications packed at less than 120°F

Specifications

Specifications

Screw Cap
Borosilicate Glass
Phenolic
Screw
Clear
Provide resistance to heat shock and chemical leaching
Pulp/Tin Foil
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit