Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Clear Borosilicate Vial with PP Hole Cap and PTFE/Silicone Septa

Offers maximum visibility

$234.00 - $511.00

Specifications

Certifications/Compliance US EPA
Closure Type Screw Cap
Color Clear
Material Borosilicate Glass
For Use With (Application) Provides resistance to heat shock and chemical leaching
View More Specs

Products 4
Catalog Number Mfr. No. Volume (Metric) Diameter (Metric) Outer Height (Metric) Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Diameter (Metric) Outer Height (Metric) Quantity Price Quantity & Availability  
02-992-144
View Documents
Qorpak™
GLC01029
40 mL 27.7 mm 95 mm 144/Cs.
Case of 144 for $473.00
Add to cart
 
02-992-757
View Documents
Qorpak™
GLC01042
40 mL 29.2 mm 81 mm 144/Cs.
Case of 144 for $511.00
Add to cart
 
02-992-841
View Documents
Qorpak™
GLC01015
20 mL 27.2 mm 57.5 mm 72/Cs.
Case of 72 for $234.00
Add to cart
 
02-992-587
View Documents
Qorpak™
GLC07508
30 mL 25 mm 95 mm 144/Cs. This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

  • Material: Type I borosilicate glass
  • Provides resistance to heat shock and chemical leaching

Specifications

Specifications

US EPA
Clear
Provides resistance to heat shock and chemical leaching
Screw Cap
Borosilicate Glass
Screw
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit