Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Clear Borosilicate Vials, Vacuum and Ionized Available on GSA/VA Contract for Federal Government customers only.

Free from contaminants such as loose dirt, carton lint, aerosols and fine glass particles

$138.00 - $364.00

Specifications

Closure Type Screw Cap
Color Clear
Material Borosilicate Glass
For Use With (Application) Free from contaminants such as loose dirt, carton lint, aerosols and fine glass particles
Grade Vacuum and ionized
View More Specs

Products 11
Catalog Number Mfr. No. Volume (Metric) Height (Metric) Closure Material Liner Material Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Height (Metric) Closure Material Liner Material Price Quantity & Availability  
02-991-468
View Documents
Qorpak™
GLC05185
4 mL 45 mm Thermoset F217/PTFE
Case of 144 for $162.00
Add to cart
 
02-991-506
View Documents
Qorpak™
GLC01010
30 mL 95 mm Phenolic Polycone
Case of 144 for $265.00
Add to cart
 
02-991-508 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Qorpak™
GLC05186
7.5 mL 60 mm Thermoset F217/PTFE
Case of 144 for $179.00
Add to cart
 
02-991-965
View Documents
Qorpak™
GLC05028
20 mL 57.5 mm Phenolic Polycone
Case of 72 for $138.00
Add to cart
 
02-992-537
View Documents
Qorpak™
GLC00996
15 mL 70 mm Phenolic Polycone
Case of 144 for $215.00
Add to cart
 
02-992-638
View Documents
Qorpak™
GLC00992
11 mL 65 mm Phenolic Polycone
Case of 144 for $237.00
Add to cart
 
02-992-775
View Documents
Qorpak™
GLC01021
40 mL 95 mm Thermoset F217/PTFE
Case of 72 for $176.00
Add to cart
 
02-992-802 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Qorpak™
GLC05027
7.5 mL 60 mm Phenolic Polycone
Case of 144 for $226.00
Add to cart
 
02-992-132
View Documents
Qorpak™
GLC07467
4 mL 45 mm Phenolic Polycone
Case of 144 for $180.00
Add to cart
 
02-992-150
View Documents
Qorpak™
GLC09126
20 mL 57.5 mm Thermoset F217/PTFE
Case of 144 for $364.00
Add to cart
 
02-992-637
View Documents
Qorpak™
GLC09161
11 mL 65 mm Thermoset F217/PTFE
Case of 144 for $238.00
Add to cart
 
Description

Description

  • Odorless and tasteless
  • Air inside vial does not interfere with contents
  • Available with caps made of different materials (Green Thermoset F217 and PTFE and black phenolic PolyCone)
  • Available in volumes ranging from 1 to 10 dr. (3.75 to 40mL)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Screw Cap
Borosilicate Glass
Vacuum and ionized
Clear
Free from contaminants such as loose dirt, carton lint, aerosols and fine glass particles
Screw
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit