Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Clear Boston Round Bottles, Cleaned for Volatiles

Offers maximum visibility and product integrity

Manufacturer:  Qorpak™ GLC01186

 View more versions of this product

Catalog No. 02-992-173


  Check Availability
Add to cart

Description

Description

  • Narrow screw neck opening
  • Curved shoulders and round body
  • Material: glass
  • Minimal chemical interference with contents
  • Environmentally sensitive
  • For long term storage

Certifications

Meets U.S. EPA Specifications and Guidelines for Contaminant-Free Sample Containers

Specifications

Specifications

16 oz., 480mL
16 oz.
F217 and PTFE
2.95 in.
Glass
Green
75mm
Screw
480mL
Thermoset
Clear
6.6 in.
12/Cs.
28-400
168mm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit