Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Clear Boston Round Bottles, Ultra Clean

Designed for those needing containers cleaned for general lab use or fluid sampling. Qorpak™ Clear Boston Round Bottles, Ultra Clean feature a proprietary cleaning process that removes up to 99.5% of particles that are 5 microns and larger.

Manufacturer:  Qorpak™ GLC01117

 View more versions of this product

Catalog No. 02-992-389


  Check Availability
Add to cart

Description

Description

  • Cleaned using three-step process
  • Made of clear glass
  • Provide maximum content visibility
  • Available with caps made of green Thermoset F217 and PTFE and black Phenolic Pulp/Vinyl
Specifications

Specifications

2 oz., 60mL
2 oz.
Pulp/Vinyl
1.53 in.
Glass
Black
39mm
Screw
60mL
Phenolic
Clear
3.7 in.
24/Cs.
20-400
94mm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit