Learn More
Qorpak™ Clear Boston Round Bottles with White Polypropylene Hole Cap and Bonded PTFE/Silicone Septa
For liquids, product storage, and field or plant sampling projects. Qorpak™ Clear Boston Round Bottles with White Polypropylene Hole Cap and Bonded PTFE/Silicone Septa help to insure product integrity for long term storage.
Manufacturer:  Qorpak™ GLC06417
 View more versions of this product
Catalog No. 02-992-019
Add to cart
Description
Description
  • Features a narrow screw neck opening, curved shoulders, and a round body
  • Glass offers minimal chemical interference with contents
  • Environmental sensitive, helps eliminate waste
Specifications
Specifications
32 oz., 960mL
32 oz.
Glass
White
960mL
Clear
12/Cs.
Screw