Learn More
Qorpak™ Clear Glass Jugs, Ultra Clean Available on GSA/VA Contract for Federal Government customers only.
For general lab use or fluid sampling. Qorpak™ Clear Glass Jugs, Ultra Clean provides content visibility.
Manufacturer:  Qorpak™ GLC01431
 View more versions of this product
Catalog No. 02-992-000
  Check Availability
Add to cart
Specifications
Description
  • Cleaning process eliminate particulate contamination
  • Available with caps made of different materials (Green Thermoset F217 and PTFE and Black Phenolic Pulp/Vinyl)
Description & Specifications
Specifications
128 oz., 3785mL
128 oz.
Thermoset
Clear
Molded finger grip
Glass
Green
166 mm
Screw
3785mL
Ultra-cleaned
F217 and PTFE
6.54 in.
11.54 in.
4/Cs.
38-400
293mm