Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Clear Glass Jugs with Black Phenolic Pulp/Tin Foil Cap Available on GSA/VA Contract for Federal Government customers only.

Storage solution for large quantities of liquid

Manufacturer:  Qorpak™ GLC01443

 View more versions of this product

Catalog No. 02-991-882


  Check Availability
Add to cart

Description

Description

  • Features molded finger grip
  • With black phenolic pulp/tin foil cap

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

128 oz., 3785mL
128 oz.
Pulp/Tin Foil
6.54 in.
11.54 in.
4/Cs.
38-400
293mm
Cylindrical
3785mL
Phenolic
Clear
Molded finger grip
Glass
Black
166 mm
Narrow
Screw
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit