Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Clear Graduated Medium Round Ready to Clean Bottles

With slight shoulder for easy pouring and handling

Manufacturer:  Qorpak™ GLC01497

 View more versions of this product

Catalog No. 02-992-158


Add to cart

Description

Description

  • Made of glass
  • Wide mouth
  • Graduated in both milliliters and ounces
  • Multipurpose containers for use with biological and pathological specimens and for storing industrial laboratory chemicals and reagents
  • With black polypropylene caps and PTFE discs
  • Available in volumes ranging from 1 to 8 oz. (30 to 240mL)

Specifications

Specifications

4 oz., 120mL
4 oz.
PTFE
2.01 in.
Glass
24/Cs.
48-400
102mm
120mL
Polypropylene
Clear
4.02 in.
Plain Carton
Black
51mm
Screw
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit