Learn More
Qorpak™ Clear Polystyrene Snap Cap Vials Available on GSA/VA Contract for Federal Government customers only.
Offers rigidity and glass-like clarity
Manufacturer:  Qorpak™ PLC03734
 View more versions of this product
Catalog No. 02-992-605
Add to cart
Description
Description
  • Caps snap on/off easily
  • For storing bench top accessories and field sampling
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
144 Case
13 dr.
35mm
33mL
Storing bench top accessories and field sampling
36mm white LDPE unlined cap
Snap Top Vial
Clear
Polystyrene
Screw cap
62mm